Sino Biological™ IL1RAPL2 (IL1R9) Recombinant Human Protein, hIgG1-Fc Tag Non-distribution product as customer accommodation.

Expressed from DNA sequence encoding extracellular domain (Met 1-Glu 356) of human IL1RAPL2 precursor fused to the Fc region of human IgG1

Manufacturer: Sino Biological™ 10156H02H5

 View more versions of this product

Catalog No. 10-156-H02H5

  Check Availability
Add to Cart

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.


    Specifications

    Common Name Interleukin-1 receptor accessory protein-like 2
    Molecular Weight 66kd
    Purity ≥90% by SDS-PAGE
    Quantity 5μg
    Source Human
    Storage Requirements −20° to −70°C
    Structural Form Recombinant
    Endotoxin Concentration <1 EU/μg
    Gene Symbol L1RAPL2
    Protein Tag hIgG1-Fc Tag
    Validated Application Assay Standard
    Protein Family Cytokines and Receptors
    Expression System Mammalian
    Product Type IL1RAPL2 (IL1R9) Recombinant Human Protein
    Format Lyophilized
    Gene ID (Entrez) 26280

    Expressed from DNA sequence encoding extracellular domain (Met 1-Glu 356) of human IL1RAPL2 precursor fused to the Fc region of human IgG1

    • Lyophilized in 100MM Glycine, 10MM NaCl , 50MM Tris , pH 7.5

  • Description & Specifications

Related Products


Product Content Feedback

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit