Learn More
Abnova™ Il2 (Mouse) ELISA Kit
Sandwich enzyme immunoassay for the quantitative measurement of mouse Il2
Manufacturer:  Abnova™ KA4810
Catalog No. 89-948-793
Add to cart
Description
Description
  • Used for the in vitro quantitative determination of mouse IL-2 in mouse serum, mouse plasma, cell lysate, culture supernatants and buffered solution
  • Assay will recognize native and recombinant mouse IL-2
Specifications
Specifications
P04351
Biotin
Quantification
Mouse IL-2 microtiter plate
For research use only (RUO)
Store the kit at 4°C.
Buffered Solution, Cell Lysates, Culture Supernatant, Plasma, Serum
3.91 to 250 pg/mL
Il2 (Mouse) ELISA Kit is a sandwich enzyme immunoassay for the quantitative measurement of mouse Il2.
16183
ELISA Kit
100 μL
Mouse
Colorimetric