Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL20RB Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA549927

Catalog No. PA549927


Add to cart

Description

Description

The antibody detects endogenous levels of total IL20RB protein.

IL20RB and IL20RA (MIM 605620) form a heterodimeric receptor for interleukin-20 (IL20; MIM 605619) (Blumberg et al., 2001 [PubMed 11163236]).[supplied by OMIM, Feb 2009].
Specifications

Specifications

IL20RB
Polyclonal
Unconjugated
IL20RB
Liquid
IL20RB
Synthesized peptide derived from internal of human IL20RB.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Western Blot
1 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Q6UXL0
DIRS1
Rabbit
IgG
100 μL
-20°C
Primary
213208, 53833
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit