Learn More
IL20RB Rabbit anti-Human, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 205211AP
Catalog No. 50-173-2933
Add to cart
Description
Description
IL20RB and IL20RA.[supplied by OMIM, Feb 2009].
Specifications
Specifications
IL20RB
0.27 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q6UXL0
DIRS1, FNDC6, IL 20 receptor subunit beta, IL 20R beta, IL 20R2, IL 20RB, IL20RB, interleukin 20 receptor beta
Rabbit
IgG
150 μL
-20°C
Primary
53833
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Western Blot
Unconjugated
IL20RB
Liquid
IL20RB
IL20RB Fusion Protein Ag14368
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human