Learn More
IL23R Mouse anti-Human, Clone: OTI5C4, lyophilized, TrueMAB™ Non-distribution product as customer accommodation.
Mouse Monoclonal Antibody
Manufacturer:  OriGeneSupplier Diversity Partner CF807388
Catalog No. 50-166-8251
Add to cart
Description
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
IL-23R is a 629 aa, novel subunit of the receptor for cytokine IL23A/IL23 which pairs with the receptor molecule IL12RB1/IL12R beta 1and thus take part in IL23A signaling. Signal transduction analysis confirms that this protein associates constitutively with Janus kinase 2 (JAK2), and also binds to transcription activator STAT3 in a ligand-dependent manner. Alternative splicing results in six spliced isoforms of IL-23R. Reports suggest that IL-23R plays an important role in psoriasis pathogenesis and is also a potential therapeutic target in inflammatory bowel disease.
Specifications
Specifications
IL23R
Monoclonal
Unconjugated
IL23R
Lyophilized
IL23R
Human recombit protein fragment corresponding to amino acids 57-355 of human IL23R produced in E.coli.
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Western Blot
OTI5C4
PBS with 8% trehalose and no preservative; pH 7.3
Q5VWK5
interleukin 23 receptor
Mouse
IgG2a
100 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
149233