Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IL23R Mouse anti-Human, Clone: OTI5H9, lyophilized, TrueMAB™
Mouse Monoclonal Antibody
Supplier: OriGene CF807494
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
IL-23R is a 629 aa, novel subunit of the receptor for cytokine IL23A/IL23 which pairs with the receptor molecule IL12RB1/IL12R beta 1and thus take part in IL23A signaling. Signal transduction analysis confirms that this protein associates constitutively with Janus kinase 2 (JAK2), and also binds to transcription activator STAT3 in a ligand-dependent manner. Alternative splicing results in six spliced isoforms of IL-23R. Reports suggest that IL-23R plays an important role in psoriasis pathogenesis and is also a potential therapeutic target in inflammatory bowel disease.Specifications
IL23R | |
Monoclonal | |
Unconjugated | |
IL23R | |
interleukin 23 receptor | |
Mouse | |
Affinity Chromatography | |
RUO | |
149233 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Western Blot | |
OTI5H9 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
Q5VWK5 | |
IL23R | |
Human recombit protein fragment corresponding to amino acids 57-355 of human IL23R produced in E.coli. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
Provide Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction