Learn More
IL24 Mouse anti-Human, Clone: OTI4H4, lyophilized, TrueMAB™ Non-distribution product as customer accommodation.
Mouse Monoclonal Antibody
Manufacturer:  OriGeneSupplier Diversity Partner CF809452
Catalog No. 50-166-9504
Add to cart
Description
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
Interleukin 24 (IL-24)/melanoma differentiation-associated gene-7 (mda-7) encodes a secreted protein that exhibits significant homology to the interleukin 10 (IL-10) family of cytokines. The human IL-24 is secreted by activated peripheral blood mononuclear cells and binds two heterodimeric receptors, IL-22R1/IL-20R2 and IL-20R1/IL-20R2 resulting in the activation of JAK/STAT signaling pathways. Mda-7/IL-24, selectively induces growth suppression, apoptosis and radiosensitization in maligt gliomas in a p53-independent manner.
Specifications
Specifications
IL24
Monoclonal
Unconjugated
IL24
Lyophilized
IL24
Human recombit protein fragment corresponding to amino acids 52-206 of human IL24 produced in E.coli.
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Immunohistochemistry (Paraffin), Western Blot
OTI4H4
PBS with 8% trehalose and no preservative; pH 7.3
Q13007
C49A, FISP, IL10B, MDA7, MOB5, ST16
Mouse
IgG1
100 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
11009