Learn More
Abnova™ IL25 (Human) ELISA Kit
For the Quantitative detection of Human IL25.
Manufacturer:  Abnova™ KA4453
Catalog No. 89-948-841
Add to cart
Description
Description
  • IL25 (Human) ELISA Kit is a sandwich enzyme immunoassay for the quantitative measurement of human IL25.
Specifications
Specifications
Human
64806
Store at 4°C for 6 months. Store at -20°C for 12 months. Avoid multiple freeze-thaw cycles.
IL25
interleukin 25
Kit
Human
1 Kit