Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL28A Mouse anti-Human, Clone:7B9A4, Abnova™

Mouse monoclonal antibody raised against recombinant human IL28A.

Manufacturer:  Abnova Corporation MAB17174

Catalog No. 89-333-597


Add to cart

Description

Description

This gene encodes a cytokine distantly related to type I interferons and the IL-10 family. This gene, interleukin 28B (IL28B), and interleukin 29 (IL29) are three closely related cytokine genes that form a cytokine gene cluster on a chromosomal region mapped to 19q13. Expression of the cytokines encoded by the three genes can be induced by viral infection. All three cytokines have been shown to interact with a heterodimeric class II cytokine receptor that consists of interleukin 10 receptor, beta (IL10RB) and interleukin 28 receptor, alpha (IL28RA). [provided by RefSeq]
Specifications

Specifications

IL28A
7B9A4
interleukin 28A (interferon, lambda 2)
In PBS (0.05% sodium azide)
Liquid
Mouse
IgG1
Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Monoclonal
Human
ELISA, Immunohistochemistry (Paraffin), Western Blot
Unconjugated
ELISA (1:10000)
Western Blot (1:500-1:2000)
Immunohistochemistry (1:200-1:1000)
Immunocytochemistry
Flow Cytometry
The optimal working dilution should be determined by the end user.
IL28A
IFNL2, IL-28A
Recombinant protein corresponding to amino acid 1-200 of human IL28A from E. coli.
100μg
Antibody
282616
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit