Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL2RA, anti-Human, Clone: Basiliximab, Abnova™

Monoclonal Antibody

Manufacturer:  Abnova Corporation RAB00048

Catalog No. 89-032-391


Add to cart

Specifications

Specifications

IL2RA
Basiliximab
Blocking Flow Cytometry (1 ug/mL) Immunofluorescence Immunohistochemistry (5 ug/mL) Western Blot (1 ug/mL) The optimal working dilution should be determined by the end user.
interleukin 2 receptor, alpha
CD25|IDDM10|IL2R|TCGFR
IgG1, kappa
RUO
Antibody
Monoclonal
This antibody recognizes the human IL2RA.
Blocking Assay, Flow Cytometry, Immunofluorescence, Immunohistochemistry, Western Blot
Unconjugated
In PBS, pH 7.2 (0.02% Proclin 300).
Liquid
IL2RA
200 μg
Store at 4°C for up to 3 months. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing.
Primary
3559
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit