Learn More
IL2RA Rabbit anti-Human, Unconjugated, Clone: AFGC-9, Abnova™
Rabbit Monoclonal Antibody.
Manufacturer:  Abnova Corporation MAB22032
Catalog No. 89-339-770
Add to cart
Description
Description
IL2RA Monoclonal antibody specifically detects IL2RA in Human samples. It is validated for IHC-P.
Specifications
Specifications
IL2RA
AFGC-9
interleukin 2 receptor, alpha
Liquid
Rabbit
IgG
Store at -20°C. Aliquot to avoid repeated freezing and thawing.
Monoclonal
Human
Immunohistochemistry (Paraffin)
Unconjugated
IL2RA
CD25, IDDM10, IL2R, TCGFR
A synthetic peptide corresponding to human IL2RA
100 μg
Primary
3559