Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IL2RG Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
Supplier: Proteintech Group Inc 114091AP150UL

Description
IL2RG is an important signaling component of many interleukin receptors, including those of interleukin -2, -4, -7 and -21, and is thus referred to as the common gamma chain. Mutations in this gene cause X-linked severe combined immunodeficiency (XSCID), as well as X-linked combined immunodeficiency (XCID), a less severe immunodeficiency disorder.Specifications
IL2RG | |
Polyclonal | |
Unconjugated | |
IL2RG | |
CD132, IL 2 receptor subunit gamma, IL 2R subunit gamma, IL 2RG, IL2RG, IMD4, p64, SCIDX, SCIDX1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
140924, 16186, 3561 | |
-20°C | |
Liquid |
Flow Cytometry, Western Blot | |
0.14 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P31785, P34902 | |
IL2RG | |
IL2RG Fusion Protein Ag1911 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction