Learn More
ACROBiosystems Recombinant Protein;Human IL-2 Protein, Tag Free;E.coli;100UG;IL2,IL2,IL-2,TCGF,lymphokine,Interleukin 2,Interleukin-2,Interleukin2
Supplier:  ACROBiosystems IL2H4113100UG
Encompass_Preferred
Recombinant Protein;Human IL-2 Protein, Tag Free;100UG;Human IL-2, Tag Free (IL2-H4113) is expressed from E.coli cells. It contains AA Ala 21 - Thr 153 (Accession # P60568).;This protein carries no "tag". The protein has a calculated MW of 15.4 kDa. The protein migrates as 17 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human IL-2, Tag Free (Cat. No. IL2-H4113) at 10μg/mL (100 μL/well) can bind Human IL-2 R beta, Fc Tag (Cat. No. ILB-H5253) with a linear range of 0.625-5 μg/mL (Routinely tested).;Immobilized Human IL-2 R alpha, Fc Tag (Cat. No. ILA-H5251) at 2μg/mL (100 μL/well) can bind Human IL-2, Tag Free (Cat. No. IL2-H4113) with a linear range of 0.2-1.56 ng/mL (QC tested).;Human IL-2, Tag Free (Cat. No. IL2-H4113) stimulates proliferation of CTLL-2 cells. The ED50 for this effect is 0.284-0.355 ng/ml (Routinely tested).
Catalog No. 50-165-7444
Item Discontinued This item has been discontinued by the supplier and is no longer available. Please reach out to your sales representative.