Learn More
Abnova™ Il34 (Mouse) ELISA Kit
For the Quantitative detection of Mouse Il34.
Manufacturer:  Abnova™ KA4496
Catalog No. 89-948-900
Add to cart
Description
Description
  • Il34 (Mouse) ELISA Kit is a sandwich enzyme immunoassay for the quantitative measurement of mouse Il34.
Specifications
Specifications
Human
76527
Store at 4°C for 6 months. Store at -20°C for 12 months. Avoid multiple freeze-thaw cycles.
Il34
interleukin 34
Kit
Mouse
1 Kit