Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL3RA Mouse anti-Human, Unconjugated, Clone: IL3RA/1531, Abnova™

Mouse Monoclonal Antibody.

Manufacturer:  Abnova Corporation MAB21257

Catalog No. 89-337-748


Add to cart

Description

Description

IL3RA Monoclonal antibody specifically detects IL3RA in Human samples. It is validated for IHC-P.
Specifications

Specifications

IL3RA
IL3RA/1531
interleukin 3 receptor, alpha (low affinity)
Liquid
Mouse
IgG2b, Kappa
Store at 4°C
Monoclonal
Human
Immunohistochemistry (Paraffin)
Unconjugated
IL3RA
CD123, IL3R, IL3RAY, IL3RX, IL3RY, MGC34174, hIL-3Ra
Recombinant protein corresponding to amino acids 26-171 of human IL3RA
100 μg
Primary
3563
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit