Learn More
IL4 293T Cell Overexpression Lysate (Denatured), Abnova
Manufacturer: Abnova Corporation H00003565T01
Description
Western Blotting
Specifications
Western Blot | |
IL4 293T Cell Transient Overexpression Lysate(Denatured) | |
16.94 | |
pCMV-IL4 full-length | |
100 μL | |
Human |
293T | |
NM_000589 | |
6.8 | |
Transient overexpression cell Lysate was tested with Anti-IL4 antibody (H00003565-B01) by western blotting | |
1X Sample Buffer (50 mM Tris-HCl, 2% SDS, 10% glycerol, 300 mM 2-mercaptoethanol, 0.01% Bromophenol blue) |