Learn More
Abnova™ IL4 (Human) ELISA Kit
Sandwich enzyme immunoassay for quantitative measurement of human IL4.
Manufacturer:  Abnova™ KA4805
Catalog No. 89-948-908
Add to cart
Description
Description
  • Used for the in vitro quantitative determination of human IL-4 in human serum, human plasma, cell lysate, culture supernatants and buffered solution
  • Assay will recognize native and recombinant human IL-4
Specifications
Specifications
P05112
1.56 to 100 pg/mL
IL4 (Human) ELISA Kit is a sandwich enzyme immunoassay for the quantitative measurement of human IL4.
3565
ELISA Kit
100 μL
Human
Colorimetric
Biotin
Quantification
Human IL-4 microtiter plate (96 wells)
For research use only (RUO)
Store the kit at 4°C.
Buffered Solution, Cell Lysates, Culture Supernatant, Plasma, Serum