Learn More
Abnova™ IL4 (Rat) ELISA Kit
Sandwich enzyme-linked immunosorbent assay
Manufacturer:  Abnova™ KA5691
Catalog No. 89-339-622
Add to cart
Description
Description
IL4 (Rat) ELISA Kit is a sandwich enzyme-linked immunosorbent assay for quantitative detection of rat IL4 in cell culture supernates and serum.
Specifications
Specifications
Quantitative
Il4
1 Kit
RUO
Rat
287287
Standard curve
Store at 4°C for 6 months, at -20°C for 12 months. Avoid multiple freeze-thaw cycles.
Il4e12