Learn More
Abnova™ IL4R 293T Cell Transient Overexpression Lysate (Denatured)
Human overexpression lysate
Manufacturer:  Abnova™ H00003566T02
Catalog No. 89-948-923
Add to cart
Description
Description
Specifications
Specifications
293T
NM_001008699
pCMV-IL4R full-length
100 uL
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Western Blot
IL4R 293T Cell Transient Overexpression Lysate(Denatured)
Theoretical M.W. 25.6kDa
Transient overexpression cell lysate was tested with anti-IL4R antibody (Mfr. No. H00003566-B01P) by Western Blot.
1X Sample Buffer (50 mM Tris-HCl, 2% SDS, 10% glycerol, 300 mM 2-mercaptoethanol, 0.01% Bromophenol blue)
Human