Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IL4R Rabbit anti-Human, Polyclonal, Abnova™
Rabbit Polyclonal Antibody
Supplier: Abnova Corporation PAB30273
Description
Rabbit polyclonal antibody raised against partial recombinant human IL4R.
- High specificity
- High reactivity
Specifications
IL4R | |
Polyclonal | |
Rabbit polyclonal antibody raised against partial recombinant human IL4R. | |
In PBS, pH 7.2 (40% glycerol, 0.02% sodium azide). | |
CD124/IL4RA | |
Rabbit | |
Antigen affinity purification | |
RUO | |
3566 | |
Store at 4°C for short term storage. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
IgG |
Immunohistochemistry (PFA fixed) | |
Unconjugated | |
Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1:50 - 1:200) The optimal working dilution should be determined by the end user. | |
P24394 | |
IL4R | |
Recombinant protein corresponding to amino acids 36-136 of human IL4R. | |
100 μL | |
Primary | |
Human | |
Liquid |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?
Provide Content Correction