Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL6 Mouse anti-Bovine, Clone: 10B2, Abnova™

Mouse monoclonal antibody raised against bovine IL6.

Manufacturer:  Abnova Corporation MAB12442

Catalog No. 89-934-373

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Specifications

Specifications

IL6
10B2
Lyophilized from PBS, pH 7.2 (0.09% sodium azide)
IL6
Recombinant protein corresponding to bovine IL6.
Antigen Affinity Purification
RUO
Primary
280826
ELISA
Unconjugated
Lyophilized
Murine
IgG2b
1mg
Store at 4°C on dry atmosphere. After reconstitution with deionized water, store at −20°C or lower. Aliquot to avoid repeated freezing and thawing.
Monoclonal
Bovine
SDS
Documents
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit