Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL6 Mouse anti-Human, Clone: 6D9A1 + 6C12B1, Abnova™

Mouse monoclonal antibody raised against human IL6.

Manufacturer:  Abnova Corporation MAB12517

Catalog No. 89-934-370


Add to cart

Description

Description

This gene encodes a cytokine that functions in inflammation and the maturation of B cells. The protein is primarily produced at sites of acute and chronic inflammation, where it is secreted into the serum and induces a transcriptional inflammatory response through interleukin 6 receptor, alpha. The functioning of this gene is implicated in a wide variety of inflammation-associated disease states, including suspectibility to diabetes mellitus and systemic juvenile rheumatoid arthritis. [provided by RefSeq]

Specifications

Specifications

IL6
6D9A1 + 6C12B1
Mouse monoclonal antibody raised against human IL6.
In ascites (0.03% sodium azide).
BSF2/HGF/HSF/IFNB2/IL-6
Mouse
IgG1
RUO
Primary
3569
ELISA, Immunohistochemistry (PFA fixed), Western Blot
Unconjugated
ELISA (1:10000) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1:200) Western Blot (1:200-1:1000) The optimal working dilution should be determined by the end user.
Liquid
IL6
Recombinant protein corresponding to human IL6.
50 uL
Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Monoclonal
Human
SDS
Documents
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit