Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IL6R Rat anti-Mouse, Clone: RM0114-14F33, Novus Biologicals™
Rat Monoclonal Antibody has been used in 1 publication
Supplier: Novus Biologicals NBP1224920.025MG
Description
IL6R Monoclonal specifically detects IL6R in Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Frozen.Specifications
IL6R | |
Monoclonal | |
LYOPH | |
Western Blot 1:500-1:1000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Frozen 1:50-1:100 | |
P22272 | |
IL6R | |
Mouse recombinant protein of the extracellular domain of IL-6 R | |
0.025 mg | |
Cancer, Immunology, Innate Immunity | |
3570 | |
Reconstitute with 0.2 ml sterilized PBS to a final concentration of 0.5 mg/ml. | |
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. | |
IgG2 |
Immunohistochemistry (Frozen), Western Blot, Immunohistochemistry | |
RM0114-14F33 | |
Unconjugated | |
A 0.2 um filtered solution in PBS with No Preservative | |
CD126, CD126 antigen, gp80, IL-6 receptor subunit alpha, IL-6R 1, IL-6R subunit alpha, IL-6R-1, IL6RA, IL-6RA, IL-6R-alpha, interleukin 6 receptor, interleukin 6 receptor alpha subunit, interleukin-6 receptor subunit alpha, Membrane glycoprotein 80, MGC104991 | |
Rat | |
Protein G purified | |
RUO | |
Primary | |
This was produced from a hybridoma (mouse myeloma fused with spleen cells from a rat immunized with mouse recombinant protein of the extracellular domain of IL-6 R. | |
Mouse | |
Purified |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only