Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IL6R Rat anti-Mouse, Clone: RM0114-14F33, Novus Biologicals™
Rat Monoclonal Antibody has been used in 1 publication
$177.16 - $493.00
Specifications
Antigen | IL6R |
---|---|
Clone | RM0114-14F33 |
Concentration | LYOPH |
Dilution | Western Blot 1:500-1:1000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Frozen 1:50-1:100 |
Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Frozen) |
Description
IL6R Monoclonal specifically detects IL6R in Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Frozen.Specifications
IL6R | |
LYOPH | |
Western Blot, Immunohistochemistry, Immunohistochemistry (Frozen) | |
Unconjugated | |
Rat | |
Cancer, Immunology, Innate Immunity | |
P22272 | |
3570 | |
Mouse recombinant protein of the extracellular domain of IL-6 R | |
Primary | |
This was produced from a hybridoma (mouse myeloma fused with spleen cells from a rat immunized with mouse recombinant protein of the extracellular domain of IL-6 R. |
RM0114-14F33 | |
Western Blot 1:500-1:1000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Frozen 1:50-1:100 | |
Monoclonal | |
Purified | |
RUO | |
Mouse | |
CD126, CD126 antigen, gp80, IL-6 receptor subunit alpha, IL-6R 1, IL-6R subunit alpha, IL-6R-1, IL6RA, IL-6RA, IL-6R-alpha, interleukin 6 receptor, interleukin 6 receptor alpha subunit, interleukin-6 receptor subunit alpha, Membrane glycoprotein 80, MGC104991 | |
IL6R | |
IgG2 | |
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
For Research Use Only
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title