Learn More
IL6R Rat anti-Mouse, Janelia Fluor 646, Clone: RM0114-14F33, Novus Biologicals™
Rat Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP122492JF646
Catalog No. NBP12492646
Add to cart
Description
Description
IL6R Monoclonal antibody specifically detects IL6R in Mouse samples. It is validated for Immunohistochemistry, Immunohistochemistry-Frozen.
Specifications
Specifications
IL6R
RM0114-14F33
Immunohistochemistry, Immunohistochemistry-Frozen
Purified
IL6R
Mouse recombinant protein of the extracellular domain of IL-6 R
Protein G purified
Cancer, Cytokine Research, Immunology, Innate Immunity
Primary
3570
Mouse
Immunohistochemistry, Immunohistochemistry (Frozen)
Janelia Fluor 646
50mM Sodium Borate with 0.05% Sodium Azide
CD126, CD126 antigen, gp80, IL-6 receptor subunit alpha, IL-6R 1, IL-6R subunit alpha, IL-6R-1, IL6RA, IL-6RA, IL-6R-alpha, interleukin 6 receptor, interleukin 6 receptor alpha subunit, interleukin-6 receptor subunit alpha, Membrane glycoprotein 80, MGC104991
Rat
IgG2
0.1mL
Store at 4°C in the dark.
Monoclonal
This was produced from a hybridoma (mouse myeloma fused with spleen cells from a rat immunized with mouse recombinant protein of the extracellular domain of IL-6 R.