Learn More
IL7R 293T Cell Overexpression Lysate (Denatured), Abnova
Manufacturer:  Abnova Corporation H00003575T01
Catalog No. 89-009-159
Add to cart
Description
Description
Western Blotting
Specifications
Specifications
293T
BC067539
6.8
Transient overexpression cell Lysate was tested with Anti-IL7R antibody (H00003575-B01) by western blotting
1X Sample Buffer (50 mM Tris-HCl, 2% SDS, 10% glycerol, 300 mM 2-mercaptoethanol, 0.01% Bromophenol blue)
Human
IL7R 293T Cell Transient Overexpression Lysate(Denatured)
50.6
pCMV-IL7R full-length
100 uL
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Western Blot