Learn More
Abnova™ IL9 (Human) ELISA Kit
Sandwich enzyme immunoassay for the quantitative measurement of human IL9
Manufacturer:  Abnova™ KA4811
Catalog No. 89-948-964
Add to cart
Description
Description
  • Used for the in vitro quantitative determination of human IL-9 in human serum, human plasma, cell lysate, culture supernatants and buffered solution
  • Assay will recognize native and recombinant human IL-9
Specifications
Specifications
P15248
Biotin
Quantification
Human IL-9 microtiter plate
For research use only (RUO)
Store the kit at 4°C.
Buffered Solution, Cell Lysates, Culture Supernatant, Plasma, Serum
1.563 to 100 pg/mL
IL9 (Human) ELISA Kit is a sandwich enzyme immunoassay for the quantitative measurement of human IL9.
3578
ELISA Kit
100 μL
Human
Colorimetric