Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL9R Chimeric Recombinant Rabbit Monoclonal Antibody (AH9R2), Invitrogen™

Rabbit Recombinant Monoclonal Antibody

Supplier:  Thermo Scientific MA548205

Encompass

Catalog No. PIMA548205


Add to Cart

Description

Description

Specificity: This antibody binds specifically to the human IL-9 receptor alpha chain.

IL9R Chimeric Recombinant Monoclonal antibody specifically detects IL9R Chimeric in Mouse samples. It is validated for Flow Cytometry, Immunocytochemistry, Immunoprecipitation, Inhibition Assays
TRUSTED_SUSTAINABILITY
Specifications

Specifications

IL9R Chimeric
Recombinant Monoclonal
1 mg/mL
PBS with 0.02% ProClin 300
P15247
Rabbit
Affinity chromatography
RUO
16198
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
IgG
Flow Cytometry, Immunocytochemistry, Immunoprecipitation, Inhibition Assays
AH9R2
Unconjugated
Il9
CD129; IL-9 receptor; Il9r; IL-9R; interleukin 9 receptor; interleukin-9 receptor
This antibody was raised by immunising rats with recombinant full-length murine IL-9.
200 μg
Primary
Mouse
Liquid
Recommendations

Recommendations

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit