Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL9R Rabbit anti-Human, Polyclonal, Invitrogen

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA551132

Catalog No. PA551132


Add to cart

Description

Description

IL9R Polyclonal Antibody detects endogenous levels of IL9R protein.

This is a receptor for interleukin-9.
Specifications

Specifications

IL9R
Polyclonal
Unconjugated
IL9R
Liquid
IL9R
Synthesized peptide derived from the Internal region of human IL-9R.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse
ELISA, Western Blot
1 mg/mL
PBS with 0.5% BSA, 50% glycerol and 0.02% sodium azide; pH 7.4
Q01113
Interleukin-9 Receptor
Rabbit
IgG
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
3581
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit