Sentinel XL™ HP System Accessory, Conversion Kit, Multi-colored

Manufacturer: ILC DOVER INC S300005P

Catalog No. 19-631-025

  Check Availability
Add to Cart

    Specifications

    For Use With (Equipment) Sentinel XL™ HP System
    Product Type Kit
    Includes Bioshield head cover, 2 HEPA cartridge and plug, breathing hose
    Certifications NIOSH TC-21C-0801
    Color Multi
    Size Universal

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit