ILC Dover PAPR Battery Assembly Available on GSA/VA Contract for Federal Government customers only.

Manufacturer: ILC DOVER INC S2003

 View more versions of this product

Catalog No. 19-631-029

  Check Availability
Add to Cart

    Specifications

    Quantity Each
    For Use With (Equipment) Sentinel xl™ hp system
    Product Type Nimh Battery Pack
    Includes Charger
    Battery Life 8 hr.
    Color Black
    Shelf Life Up to 500 charge/discharge cycle

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit