Learn More
ILC Dover Sentinel XL™ HP PAPR
Tailored for infectious control and pandemic preparedness support.
Manufacturer:  ILC Dover S300011
Catalog No. 19-631-005
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
Features a one size, conformal headpiece designed to securely integrate with a single state-of-the-art blower platform adaptable to multiple applications
  • System is convertible to CBRN applications through change of head cover/hose and filters
  • Tailored head cover design permits stethoscope use while wearing
  • Secure threaded hose/hood head cover interface is easy to use
  • Audible/visual low voltage alarm— flashing LED and buzzer at 30 minutes remaining battery life
  • NiMH battery elimate memory effect associated with NiCd batteries
  • Rugged dual chamber blower design for increased durability
  • Latex free material eliminates potential for allergic reactions
    • Components
      • Blower with battery
      • HEPA filters and plug
      • Head Cover (white or premium)
      • Blower hose
      • Battery charger (for rechargeable systems)
      • Flow meter
      • User's manual
      • Quick-Loc™ Belt
Specifications
Specifications
NIOSH TC-21C-0848, TC-23C-2537
Universal
Belt
Papr System
Blower, nimh rechargeable battery and clear hood in duffle bag, quick-loc waist belt, smart battery charger, flow meter, 2 hepa cartridge, hp breathing hose