Learn More
Laxco™ iLED Series LED Light Source PROMO
Laxco™ iLED Series boost bright LED illumination in small, compact, small foot print system.
Manufacturer:  Laxco™ AMPSILED21P
Catalog No. AMPSILED21P
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Description
Description
  • 12 LED strip creates a homogenized transmitted illumination
  • Dual directional upper light guides produce 720 cd of focused LED illumination comparable to a 150 w fiber optic light source
  • Independent variable intensity controls for both upper and lower illumination
  • 50,000 hour bulb life
  • Weighted base for stand-alone option
Specifications
Specifications
Manufacturing, Research, Laboratory
110 to 220 V Auto Switch, External, 5 V/2 A
Power supply, wieghted base, light guide
Weighted Base
32.2 x 17.1 x 35.5 cm (12.6 x 6.7 x 13.9 in.)
150 w equivalent LED