Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ILF2 Mouse anti-Human, Clone: 7C5, Novus Biologicals™

Mouse Monoclonal Antibody

$116.82 - $343.85

Specifications

Antigen ILF2
Clone 7C5
Immunogen The immunogen for this antibody is ILF2.
Host Species Mouse
Primary or Secondary Primary
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP17918920 View Documents Novus Biologicals
NBP17918920UL
20ul Each for $116.82
Add to cart
 
NBP179189 View Documents Novus Biologicals
NBP179189
100ul Each for $343.85
Add to cart
 
Description

Description

ILF2 Monoclonal antibody specifically detects Antigen in Human samples. It is validated for Western Blotting.
Specifications

Specifications

ILF2
The immunogen for this antibody is ILF2.
Primary
IgM Kappa
interleukin enhancer binding factor 2, 45kD, interleukin enhancer binding factor 2, 45kDa, interleukin enhancer-binding factor 2, MGC8391, NF45PRO3063, Nuclear factor of activated T-cells 45 kDa, nuclear factor of activated T-cells, 45-kDa
PBS and 2% Sucrose with 0.09% Sodium Azide
Protein A or G purified
Store at -20C. Avoid freeze-thaw cycles.
7C5
Mouse
Monoclonal
Unconjugated
3608
Purified
Western Blot
RUO
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit