Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ILK Mouse anti-Human, Alexa Fluor™ 405, Clone: OTI2H3, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP271037AF405

Catalog No. N271037A405


Add to cart

Description

Description

ILK Monoclonal antibody specifically detects ILK in Human samples. It is validated for Western Blot.
Specifications

Specifications

ILK
OTI2H3
Western Blot
Affinity Purified, Conjugated
ILK
Recombinant protein fragment corresponding to amino acids 1-244 of human ILK (NP_004508) produced in E.coli.
Immunogen affinity purified
Phospho Specific, Protein Kinase, Signal Transduction
Primary
3611.0
Western Blot
Alexa Fluor 405
50mM Sodium Borate with 0.05% Sodium Azide
59 kDa serine/threonine-protein kinase, DKFZp686F1765, EC 2.7.11.1, ILK1, ILK-1, ILK2, ILK-2, integrin-linked kinase, integrin-linked kinase-2, integrin-linked protein kinase, P59, p59ILK
Mouse
IgG2b
0.1 mL
Store at 4°C in the dark.
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit