Learn More
Cole-Parmer Cole-Parmer Horizontal Fiber Optic Illuminator  DFS Item
Manufacturer:  Cole-Parmer 0979000
Our fan-cooled illuminator is ideal for microscopy and circuit board inspections. Transformer is attached to power cord outside the illuminator housing for cooler operating temperatures and longer bulb life. Solid-state intensity control lets you operate the illuminator in economy, medium, or high lamp settings. Order polarizer or a filter kit for special lighting effects.
Catalog No. 50-628-564