Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva illustra ExoProStar 1-Step PROMO

Cytiva illustra ExoProStar 1-Step PROMO remove unincorporated primers and dNTPs

Specifications

For Use With (Application) Enzymatic PCR clean up
Format 1 Tube
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
45-200-501LR0
View Documents
Cytiva
29489261
100 Reactions This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. View substitute products »
45-200-502LR0
View Documents
Cytiva
29489262
500 Reactions This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. View substitute products »
Description

Description

illustra ExoProStar is our new name for this PCR clean-up technology. No change in the formulation, so you get the same great product, and same great benefits!

The ExoProStar 1-Step PCR and Sequence Reaction Clean-Up reagent provides a simple, single-step method for removing unincorporated primers and nucleotides prior to downstream analysis. illustra Exonuclease I and Alkaline Phosphatase enzymes are combined in a single, stable mix requiring just one pipetting step to set up the clean-up reaction.

illustra ExoProStar 1-Step is based on GE™ founding patents for the clean-up of PCR reactions using Exonuclease 1 and Alkaline Phosphatase. This 1-step version is not available in USA, Canada and Japan. Instead, we offer a version with the enzymes in two separate tubes, the 29002090, and the faster version, 29046321.

Specifications

Specifications

Enzymatic PCR clean up
1 Tube
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit