Learn More
Cytiva illustra™ ExoProStar™ S PCR and Sequence Reaction Clean-up Kit
Enzymatic PCR clean up technology with Exonuclease I and Shrimp Alkaline Phosphatase to remove unincorporated primers and dNTPs
$22.50 - $1292.00
Specifications
For Use With (Application) PCR and Sequence Reaction;Removes unincorporated primers and dNTPs; Improves digestion efficiency; No loss of PCR product
Product Type illustra ExoProStar S
View More Specs
Products 4
Catalog Number Mfr. No. No. of Reactions Price Quantity & Availability  
Catalog Number Mfr. No. No. of Reactions Price Quantity & Availability  
45-002-350
View Documents Promotion Details
Cytiva
US79002
20
Each for $22.50
Add to cart
45-002-352
View Documents Promotion Details
Cytiva
US79050
500
Each for $394.00
Add to cart
45-002-351
View Documents Promotion Details
Cytiva
US79010
100
Each for $105.80
Add to cart
45-002-353
View Documents Promotion Details
Cytiva
US79200
2000
Each for $1,292.00
Add to cart
Description
Description
  • Easy to automate
  • Enzymes provided in two separate tubes; just two simple pipetting steps are needed to prepare the reaction
  • Enzymes optimized to work together for high efficiency removal of unincorporated primers and nucleotides
  • Scalable for different reaction sizes
  • No loss of PCR product
  • Complete heat inactivation of the enzymes within 10 minutes
  • Fast 15 minute protocol
  • Updates existing enzymatic PCR clean up technology with Exonuclease I and Shrimp Alkaline Phosphatase to remove unincorporated primers and dNTPs
  • Improves digestion efficiency with no degradation of the target PCR product
  • Sample is completely intact and ready for immediate use in manual or automated processes
Specifications
Specifications
PCR and Sequence Reaction;Removes unincorporated primers and dNTPs; Improves digestion efficiency; No loss of PCR product
illustra ExoProStar S