Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva illustra™ GenomiPhi V2 DNA Amplification Kit PROMO

Amplify whole genomes with high efficiency and representation. Cytiva Life Sciences™ illustra™ GenomiPhi V2 DNA Amplification Kit provides whole genome amplification with yields of 4 to 7μg.

$215.00 - $595.00

Specifications

For Use With (Application) Genomic DNA Amplification
Description Illustra GenomiPhi V2 DNA Amplification Kit offers highly efficient and representative whole-genome amplification with 4 to 7 μg yield from nanogram amounts of DNA sample.
Product Type illustra GenomiPhi V2 DNA Amplification Kit
Size > 10 kb
Storage Requirements Store the kit at -70°C.The enzyme mix must be stored at -70°C; all other components may be stored at -20°C. Thaw components on ice and maintain at 0°C to 4°C during handling.
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
45-001-221LR1
View Documents
Cytiva
29648019
25 Reactions
Each for $215.00
Add to cart
 
45-001-222LR1
View Documents
Cytiva
29648020
100 Reactions
Each for $595.00
Add to cart
 
Description

Description

  • Starting material can be purified DNA or nonpurified cell lysates
  • Simple protocol provides typical yields of 4 to 7μg DNA and average product lengths >10kb
  • Less hands-on time compared to traditional isolation methods
  • Representative amplification of the whole genome
  • Automation-friendly protocol with great reproducibility
  • High-quality DNA for array CGH, high-throughput genotyping, DNA archival, PCR, restriction enzyme digestion, hybridization, and cloning
  • Contains all of the components necessary for genomic DNA preparation by isothermal strand displacement amplification

Specifications

Specifications

Genomic DNA Amplification
illustra GenomiPhi V2 DNA Amplification Kit
Store the kit at -70°C.The enzyme mix must be stored at -70°C; all other components may be stored at -20°C. Thaw components on ice and maintain at 0°C to 4°C during handling.
Illustra GenomiPhi V2 DNA Amplification Kit offers highly efficient and representative whole-genome amplification with 4 to 7 μg yield from nanogram amounts of DNA sample.
> 10 kb
1.5 to 2 hr.
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit