Learn More
GE Healthcare illustra™ GFX™ PCR DNA and Gel Band Purification Kit : PROMOTION
Promotion Details: Receive a 31% discount.
Suitable for isolation and concentration of DNA fragments (50 bp to 10kb) from PCR mixtures, DNA-containing agarose gel bands, enzyme-based DNA modifications, and restriction digestions
Manufacturer:  GE Healthcare 28903471PM X
Catalog No. 45-001-490PM
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Description
Description
Typical recoveries range from 60 to 80% for DNA fragments from agarose gel to as high as 95% for PCR products from solution.
  • Rapid and easy-to-use purification method with less than 10 minutes hands-on time
  • Flexible 10 to 50µL elution volume for different DNA concentration needs
  • Improved column design with increased maximum volume holding of up to 800µL liquid volume (950µL without a cap)
  • DNA of exceptional purity (to 99.5%) is ready for direct use in sequencing, PCR, labeling, restriction enzyme digestion, and cloning
  • Visual color indicator added to the capture buffer to ensure optimal pH for maximum DNA binding and recovery
  • Kit combines a versatile chaotropic buffer with a glass fiber matrix supported in a spin-column for the purification of DNA from both solution and agarose gel
Specifications
Specifications
Isolation and concentration of DNA fragments (50bp to 10kb) from PCR mixtures, DNA-containing Agarose Gel bands, Enzyme-based DNA modifications and restriction digestions
Tris HCl and sterile Water
GFX™ columns, collection tubes, color-coded bottles of capture Buffer, Wash Buffer, two Elution Buffers
250 Preps