GE Healthcare illustra PuRe Taq Ready-To-Go™ PCR Beads - Promo

Special Offer!  (Expires 12/31/2015)
Ambient temperature stable, pre-mixed, pre-dispensed reactions optimized for performing standard PCR amplifications

Manufacturer: GE Healthcare 29168098

 View more versions of this product

Catalog No. 29-168-098

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


Includes: Room temperature stable beads containing ~ 2.5 units of recombinant PuReTaq DNA Polymerase, dATP, dCTP, dGTP, dTTP, stabilizers, BSA and reaction Buffer, When a bead is reconstituted to a 25Glc muL final volume, the concentration of each dNTP

    Specifications

    For Use With (Application) PCR application
    Quantity 1 Kit
    Storage Requirements Ambient temperature in pouch with dessicant
    Volume 200μL
    Color White
    Format 0.2mL tube/plate
    Includes Room temperature stable beads containing ~ 2.5 units of recombinant PuReTaq DNA Polymerase, dATP, dCTP, dGTP, dTTP, stabilizers, BSA and reaction Buffer, When a bead is reconstituted to a 25Glc muL final volume, the concentration of each dNTP

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit