Learn More
GE Healthcare illustra PuRe Taq Ready-To-Go™ PCR Beads - Promo
Special Offer!  (Expires 12/31/2015)
Ambient temperature stable, pre-mixed, pre-dispensed reactions optimized for performing standard PCR amplifications
Manufacturer:  GE Healthcare 29168097
 View more versions of this product
Catalog No. 29-168-097
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Includes: Room temperature stable beads containing ~ 2.5 units of recombinant PuReTaq DNA Polymerase, dATP, dCTP, dGTP, dTTP, stabilizers, BSA and reaction Buffer, When a bead is reconstituted to a 25Glc muL final volume, the concentration of each dNTP
Specifications
Description
Recommended Storage
Ambient temperature in pouch with dessicant
Description & Specifications
Specifications
White
0.2mL tube/plate
1 Kit
200μL
PCR application
Room temperature stable beads containing ~ 2.5 units of recombinant PuReTaq DNA Polymerase, dATP, dCTP, dGTP, dTTP, stabilizers, BSA and reaction Buffer, When a bead is reconstituted to a 25Glc muL final volume, the concentration of each dNTP
Ambient temperature in pouch with dessicant