Learn More
GE Healthcare illustra™ PuReTaq Ready-To-Go™ PCR Beads: PROMO
Pre-mixed, pre-dispensed reaction beads optimized for performing standard PCR amplifications.  GE Healthcare Lifescience™ illustra™ PuReTaq Ready-To-Go™ PCR Beads are stable at ambient temperature.
Manufacturer:  GE Healthcare 27955701PM
Catalog No. 46-001-011LR
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Description
Description
  • puReTaq DNA polymerase and high-purity reagents provide robust performance with very low levels of contaminating DNA
  • Manufacturing process locks in performance for excellent reproducibility with less risk of pipetting errors and contamination
  • Long-term ambient-temperature stability simplifies shipping and removes any need for freezer storage
  • Verified for real-time PCR
  • Ready-To-Go means simply add template DNA and primers and cycle
Specifications
Specifications
White
0.2mL Tubes/plate
96.0
Ambient temperature in pouch with desiccant
For PCR application
Room temperature stable beads containing ∼ 2.5 units of recombinant PuReTaq DNA Polymerase, dATP, dCTP, dGTP, dTTP, stabilizers, BSA and reaction Buffer, When a bead is reconstituted to a 25μL final volume, the concentration of each dNTP
1 Kit
200μL