Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva illustra™ PuReTaq Ready-To-Go™ PCR Beads PROMO

Cytiva illustra™ PuReTaq Ready-To-Go™ PCR Beads PROMO optimized for performing standard PCR amplifications

Specifications

Color White
For Use With (Application) PCR
Includes Room Temperature Stable Beads, Recombinant PuReTaq DNA Polymerase, dATP, dCTP, dGTP, dTTP, Stabilizers, BSA, and Reaction Buffer
Quantity 1 Kit
Storage Requirements Ambient Temperature in Pouch with Desiccant
View More Specs

Products 2
Catalog Number Mfr. No. Volume Price Quantity & Availability  
Catalog Number Mfr. No. Volume Price Quantity & Availability  
46-001-013LR0
View Documents
Cytiva
29489240
500 μL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. View substitute products »
46-001-014LR0
View Documents
Cytiva
29489241
200 μL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. View substitute products »
Description

Description

  • puReTaq DNA polymerase and high-purity reagents provide robust performance with very low levels of contaminating DNA
  • Manufacturing process locks in performance for excellent reproducibility with less risk of pipetting errors and contamination
  • Long-term ambient-temperature stability simplifies shipping and removes any need for freezer storage
  • Verified for real-time PCR
  • Ready-To-Go means simply add template DNA and primers and cycle

Specifications

Specifications

White
Room Temperature Stable Beads, Recombinant PuReTaq DNA Polymerase, dATP, dCTP, dGTP, dTTP, Stabilizers, BSA, and Reaction Buffer
Ambient Temperature in Pouch with Desiccant
PCR
1 Kit
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit