Learn More
GE Healthcare illustra™ Ready-To-Go™ GenomiPhi V3 DNA Amplification Kit
Special Offer! (Expires 12/31/2015)
Manufacturer:  GE Healthcare 29168048
 View more versions of this product
Catalog No. 29-168-048
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications Description & Specifications
Specifications
Whole genome amplification by isothermal strand displacement amplification
96 Reactions