Learn More
GE Healthcare illustra™ RNAspin Mini Isolation Kit: PROMO
Extract high-quality RNA from a wide range of samples rapidly. GE Healthcare Life Sciences™illustra™ RNAspin Mini Isolation Kit can be used to isolate total RNA from cultured cells, tissue, bacteria, or yeast.
Manufacturer:  GE Healthcare 25050072PM
Catalog No. 45-001-163LR
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Includes: Protocol booklet with detachable, quick reference card, necessary components for specified number of preps, including prefilters and DNase I
Description
Description
  • Recovers total RNA due to removal of genomic DNA through on-column DNase I treatment
  • Can be used on cultured cells, tissue, yeast and hard-to-lyse bacterial strains
  • Prefilters reduce lysate viscosity for maximum yield and purity
  • Results can be obtained with small amounts of sample
  • Binding capacity: 100μg
  • Sample size: 30mg (animal tissue), up to 5 × 106 cells (cell culture)
  • Elution volume: 40 to 120μL
  • RNA Purity (A260/A280): 1.8 to 2.2
  • Total time of preparation: <30 min/6 preps
Quantitative reverse transcriptase PCR (QRT-PCR), Northern blot, and microarray experiments
Specifications
Specifications
Rapid extraction of high-quality RNA from a wide range of samples
RNAspin Mini Isolation Kit
30mg (animal tissue), up to 5 × 106 cells (cell culture)
Protocol booklet with detachable, quick-reference guide and necessary components, including prefilters and DNase I
250 Preps
Up to 100μg