Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Our pricing system is unavailable. You are viewing list price.
Please call 1-800-766-7000 to place your order or try our site again later.

GE Healthcare illustra™ Shrimp Alkaline Phosphatase: PROMO

Product

Manufacturer:  GE Healthcare E70092YPM

Catalog No. 45-500-139LR

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Includes: 10x reaction buffer: 200mM Tris-HCl, pH 8.0, 100mM MgCl2; Dilution buffer: 50mM Tris-HCl, pH 8.0

Description

Description

  • Bacterial
  • Expressed in E.Coli
  • Inactivated by heating at 75°C for 5 minutes
  • Active in PCR buffers and restriction endonuclease buffers
  • Activity: 1U/μL
  • Purified to homogeneity
  • Free of all contaminating endonucleases, exonucleases and ribonucleases
Specifications

Specifications

Alkaline Phosphatase
Bacteria
Purified to homogeneity
500U
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit