Learn More
GE Healthcare illustra™ Single Cell GenomiPhi™ DNA Amplification Kit
Special Offer! (Expires 12/31/2015)
Manufacturer:  GE Healthcare 29168051
 View more versions of this product
Catalog No. 29-168-051
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications
Description
Recommended Storage
Store the kit at -70°C. The enzyme mix must be stored at -70°C; all other components may be stored at -20°C.
Description & Specifications
Specifications
GE Healthcare illustra Single Cell GenomiPhi DNA Amplification Kit
illustra Single Cell GenomiPhi DNA Amplification Kit
Store the kit at -70°C. The enzyme mix must be stored at -70°C; all other components may be stored at -20°C.
Single Cell GenomiPhi Reaction buffer 1 x 275µL, Single Cell GenomiPhi Enzyme mix 1 x 25µL, Single Cell GenomiPhi Amplification mix 1 x 25µL, Single Cell GenomiPhi DTT Solution 1 x 100µL, Single Cell GenomiPhi Lysis buffer 1 x 100µL and Single Cell GenomiPhi Neutralisation buffer 1 x 100µL
100 Reactions