Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GE Healthcare illustra™ Single Cell GenomiPhi™ DNA Amplification Kit: PROMO

Amplify whole genome DNA from as little as a single cell.  GE Healthcare Lifescience™ illustra™ Single Cell GenomiPhi™ DNA Amplification Kit combines unique capabilities of Phi29 DNA polymerase with an optimized formulation to amplify genomic DNA from 1-1000 cells without background interference.

Specifications

Final Product Type illustra™ Single Cell GenomiPhi™ DNA Amplification Kit
Includes Single Cell GenomiPhi Reaction buffer 1 x 275μL, Single Cell GenomiPhi Enzyme mix 1 x 25μL, Single Cell GenomiPhi Amplification mix 1 x 25μL, Single Cell GenomiPhi DTT Solution 1 x 100μL, Single Cell GenomiPhi Lysis buffer 1 x 100μL and Single Cell GenomiPhi Neutralisation buffer 1 x 100μL
Storage Requirements Store the kit at -70°C. The enzyme mix must be stored at -70°C; all other components may be stored at -20°C
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
45-500-135LR View Documents GE Healthcare
29108039PM
100 Reactions This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Description

Description

  • For whole genome amplification of genomic DNA from as little as a single cell
  • Novel clean-up process supresses background allowing sensitivity down to 1fg of input DNA
  • Simple Isothernal strand displacement (MDA) technology using Phi29 DNA polymerase
  • Amplifications can be completed in less than 3 hours with less than 15 minutes hands-on time
  • Optimized to promote complete cell lysis with uniform denaturation of DNA prior to amplification
  • Reaction time: 2 to 4 hours
  • Product average size: >10kb
Applications

For SNP arrays, Next Gen Sequencing, aCGH

Recommended Storage

Store the kit at -70°C. The enzyme mix must be stored at -70°C; all other components may be stored at -20°C


Specifications

Specifications

illustra™ Single Cell GenomiPhi™ DNA Amplification Kit
Single Cell GenomiPhi Reaction buffer 1 x 275μL, Single Cell GenomiPhi Enzyme mix 1 x 25μL, Single Cell GenomiPhi Amplification mix 1 x 25μL, Single Cell GenomiPhi DTT Solution 1 x 100μL, Single Cell GenomiPhi Lysis buffer 1 x 100μL and Single Cell GenomiPhi Neutralisation buffer 1 x 100μL
Store the kit at -70°C. The enzyme mix must be stored at -70°C; all other components may be stored at -20°C
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit