Learn More
GE Healthcare illustra™ Solution dNTPs: PCR Nucleotide Mix: PROMO
Family
Manufacturer:  GE Healthcare 28406564PM
Catalog No. 45-001-461LR
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Description
Description
  • Greater than 99% triphosphate purity (by HPLC) for optimum performance and consistency
  • Buffer-free and ready-to-use solutions in multiple formats
  • Functionally tested for long PCR and sequencing
Order Info
Custom formulation, testing, packaging, and labeling are available
Specifications
Specifications
10mM
Liquid
PCR Nucleotide Mix dNTP Mix (10mM each A,C,G,T)
500μm
Colorless
Functionally tested for long PCR and sequencing, Nucleic Acid amplification, labelling, mutagenesis, cDNA synthesis and expression profiling
>99%